Learn More
Selleck Chemical LLCSupplier Diversity Partner Rosuvastatin Calcium 100mg 147098-20-2 ZD4522  DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S2169100MG
Rosuvastatin Calcium (ZD4522) is a competitive inhibitor of HMG-CoA reductase with IC50 of 11 nM in a cell-free assay.
Catalog No. 50-665-7