Learn More
BUCHI Rotavapor™ R-300 Rotary Evaporator with Controller
The R-300 meets the highest expectations in convenience and versatility. The I-300 interface provides a central location to regulate each evaporation parameter.
Manufacturer:  BUCHI 11R300252V122PM
 View more versions of this product
Catalog No. 05-000-979PM
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
The R-300 Rotavapor™ has an ergonomic handle for easy positioning of the evaporation flask in to and out of the water bath. The patented multipurpose Combi-clip with snap lock-mechanism provides for quick flask connection and detachment with one hand.
The I-300 interface is the central control unit for all process parameters of the Rotavapor R-300. This includes rotation speed; lift position; heating, cooling, and vapor temperature; and pressure control. Push notifications on mobile devices allows connected users to view the status of multiple R-300s.
  • Intuitive user interaction
  • Precise operation control
  • 1L or 5L flask capacity
  • Ergonomic handle
  • Combi-Clip
  • Valve unit available for non-BUCHI vacuum source
  • Additional glass assemblies and configurations
Specifications
Specifications
Rotary Evaporator
R-300
CE/CSA
5 L
Electronic Lift
1600 w
Vertical
I-300 Interface
50/60 Hz
24/40
100/120 V
10 to 280 rpm
220°C
100/120 V
607 x 429 x 947 mm
1100 w