Learn More
BUCHI Rotavapor™ R-300 Rotary Evaporator with Controller and V-300 Pump
GSA_VA
The R-300 meets the highest expectations in convenience and versatility. Its modular design allows extension of the Rotavapor™ R-300 to a fully integrated system where a central interface regulates each component.
Manufacturer:  BUCHI 11SR300152VS02
 View more versions of this product
Catalog No. 05-001-028
Add to cart
Description
Description
  • Intuitive user interaction
  • Precise operation control for repeatability and safety
  • 1 L or 5 L flask capacity baths with cordless changes and cleaning
  • Up to 220 °C bath heating with 5 L bath
  • Ergonomic handle with manual or electronic lift
  • Choice of P+G coating for added protection of condenser and receiving flask
  • Unattended operation and mobile monitoring available
  • Modular system for control, vacuum, and chiller integration
  • Additional glass assemblies and configurations
  • Cloud services for optimized productivity and automated processes
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Manual Lift
100/120 V
10 to 280 rpm
220°C
1600 W
Vertical
I-300 Interface
CE/CSA
50/60 Hz
24/40
5 mbar
5 L
Woulff Bottle
V-300 Diaphragm
100/120 V
607 x 429 x 947 mm
1100 W
Rotary Evaporator System
R-300/V-300