Learn More
BUCHI Rotavapor™ R-300 Rotary Evaporator with Controller and V-300 Pump PROMO
The R-300 meets the highest expectations in convenience and versatility. Included is the V-300 vacuum pump, providing quiet vacuum over the course of your evaporation.
Manufacturer:  BUCHI 11SR300152VS02PMO
 View more versions of this product
Catalog No. 05001028PMO
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
Description
Description
The R-300 Rotavapor™ has an ergonomic handle for easy positioning of the evaporation flask in to and out of the water bath. The I-300 interface is the central control unit for all process parameters of the Rotavapor R-300. This includes rotation speed; lift position; heating, cooling, and vapor temperature; and pressure control. The I-300 interface also controls the V-300 vacuum pump and chiller (if applicable), providing reliable performance throughout the evaporation. The patented multipurpose Combi-clip with snap lock-mechanism provides for quick flask connection and detachment with one hand.
  • Intuitive user interaction
  • Precise operation control
  • 5L flask capacity
  • Ergonomic handle
  • Combi-Clip
  • Additional glass assemblies and configurations
Specifications
Specifications
5 mbar
5 L
V-300 Vacuum Pump, Manual Lift
V-300 Diaphragm
100/120 V
607 x 429 x 947 mm
1100 w
Rotary Evaporator System
R-300/V-300
100/120 V
10 to 280 rpm
220°C
1600 w
Vertical
I-300 Interface
CE/CSA
50/60 Hz
24/40