Learn More
BUCHI Rotavapor™ R-300 Rotary Evaporator Standalone
The R-300 meets the highest expectations in convenience and versatility.
Manufacturer:  BUCHI 11R300112V102PM
 View more versions of this product
Catalog No. 05-000-329PM
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
The R-300 Rotavapor™ has an ergonomic handle for easy positioning of the evaporation flask into and out of the water bath. Rotation speed, lift position and heating are controlled directly on the unit. The patented multipurpose Combi-clip with snap-lock mechanism provides for quick flask connection and detachment with one hand.
  • Intuitive user interaction
  • Precise operation control
  • 1 L flask capacity
  • Ergonomic handle
  • Combi-Clip
  • Additional glass assemblies and configurations
Specifications
Specifications
Rotary Evaporator
R-300
CE/CSA
1 L
95°C
100/120 V
607 x 429 x 947 mm
50/60 Hz
24/40
100/120 V
10 to 280 rpm
1600 w
Vertical
1100 w