Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rotavirus A MAb Mouse anti-All, Unconjugated, Clone: Clone 1, MilliporeSigma™
Mouse Monoclonal Antibody
$791.00 - $7907.00
Specifications
Antigen | Rotavirus A MAb |
---|---|
Clone | Clone 1 |
Applications | ELISA, Immunodiffusion, Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABX84511MI
|
MilliporeSigma
MABX84511KC |
1 mg |
Each for $791.00
|
|
|||||
MABX84110MI
|
MilliporeSigma
MABX845110KC |
10 mg |
Each for $7,907.00
|
|
|||||
Description
Rotavirus A MAb Monoclonal antibody specifically detects Rotavirus A MAb in All samples. It is validated for Enzyme Immunoassay (ELISA), Immunodiffusion, Western Blotting.Specifications
Rotavirus A MAb | |
ELISA, Immunodiffusion, Western Blot | |
Unconjugated | |
Infectious Diseases | |
Anti-Rotavirus | |
IgG2b | |
Affinity Purified | |
Specific for intact virus of strains: Rhesus Rotavirus, human rotavirus WA strain, SA11, etc. |
Clone 1 | |
Monoclonal | |
Mouse | |
All species | |
Viral Lysate | |
Primary | |
2 to 8°C. For long-term storage, freeze at -20°C. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title