Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rotavirus Mouse anti-Virus, Clone: CF306, Abnova™
Mouse Monoclonal Antibody
Supplier: Abnova Corporation MAB13641
Description
Mouse monoclonal antibody raised against rotavirus.
- High specificity
- High reactivity
Specifications
Rotavirus | |
Monoclonal | |
Unconjugated | |
Lateral-Flow Immunochromatographic Assay Western Blot The optimal working dilution should be determined by the end user. | |
Mouse | |
Protein A purification | |
RUO | |
This antibody reacts with p42 antigen of rotavirus. | |
Store at 4°C. | |
IgG2a |
Lateral Flow, Western Blot | |
CF306 | |
Mouse monoclonal antibody raised against rotavirus. | |
In PBS, pH 7.4 (0.09% sodium azide). | |
Bovine rotavirus. | |
1 mg | |
Primary | |
Virus | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?
Provide Content Correction