Learn More
Eppendorf™ Rotor FA-45-48-11 Available on GSA/VA Contract for Federal Government customers only.
Manufacturer:  Eppendorf™ 5427754008
Catalog No. 14-280-104
Add to cart
Description
Description
  • Max. speed: 18,210 × g (12,700 rpm)
  • Max. capacity: 48 × 1.5/2.0 mL tubes
  • Aerosol-tight for safe centrifugation of hazardous samples
  • Eppendorf QuickLock™ lid
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Fixed Angle Rotor