Learn More
Guardian Manufacturing CP-25R, Rough Finish 25 mil Butyl Gloves
Guardian™ Rough Finish 25 mil Butyl Gloves features a unique solution-dipped process that provides seamless molecular structures, which is the key to chemical impermeability.
Manufacturer:  Guardian Manufacturing 52702
 View more versions of this product
Catalog No. 19-321-130
Add to cart
Description
Description
  • Constructed of Butyl
  • Rolled Cuff
  • Unlined
  • Rough Finish
Specifications
Specifications
25 mil
Beaded/rolled
Butyl
1 pair
Black
ISO 9001, USW, USA MADE, Hubzone certified, BERRY, ASTM D412
Rough
Chemicals and gases
Small
Powdered
14 in.