Learn More
Essendant Round BRUTE™ Lids
All-plastic seamless construction
Manufacturer:  Essendant RCP2619YEL
 View more versions of this product
Catalog No. 19-046-684
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Easy to clean; withstands bumps and kicks; will not rust, chip or peel
  • Reinforced rims add strength and durability
  • Built-in handles aid lifting and allow easy, no-jam nesting
  • Double-ribbed base ensures stability and increased dragging capacity
  • Snap-on lids (sold separately) contain odors and allow secure, stable stacking
Order Info
Universal dolly for all sizes of containers available separately (19-046-694).
Certifications
Gray, white, and yellow containers are USDA Meat and Poultry Equipment Group Listed
Compliance
Assist in complying with HACCP guidelines.
Specifications
Specifications
NSF, USDA, HACCP
19.9 in.
20 gal. Brute (2620) containers
4.6 cm
Yellow
Safety
Snap-on
50.5 cm
1.8 in.
Round
Polyethylene
Lid