Learn More
Essendant Round BRUTE™ Lids
All-plastic seamless construction
Manufacturer:  Essendant FG263100RED
 View more versions of this product
Catalog No. 19-046-688
Add to cart
Description
Description
  • Easy to clean; withstands bumps and kicks; will not rust, chip or peel
  • Reinforced rims add strength and durability
  • Built-in handles aid lifting and allow easy, no-jam nesting
  • Double-ribbed base ensures stability and increased dragging capacity
  • Snap-on lids (sold separately) contain odors and allow secure, stable stacking
Order Info
Universal dolly for all sizes of containers available separately (19-046-694).
Certifications
Gray, white, and yellow containers are USDA Meat and Poultry Equipment Group Listed
Compliance
Assist in complying with HACCP guidelines.
Specifications
Specifications
NSF, USDA, HACCP
22 in.
32 gal. Brute (2632) containers
4.06 cm
Red
Safety
Snap-on
55.88 cm
1.6 in.
Built in handles, reinforced rim
LDPE
Lid