Roxindole hydrochloride, Tocris

Dopamine D2 autoreceptor agonist; Also has affinity for D3, D4, 5-HT1A receptors and the 5-HT transporter

Manufacturer: R&D Systems 1559/50

Quantity 50mg

 View more versions of this product

Catalog No. 15-595-0

  Check Availability
Add to Cart

    Chemical Identifiers

    Assay Percent Range >98%
    Molecular Formula C23H26N2O·HCl
    Formula Weight 382.93
    Synonym 3-[4-(3, 6-Dihydro-4-phenyl-1(2H)-pyridinyl)butyl]-1H-indol-5-ol hydrochloride
    Chemical Name or Material Roxindole hydrochloride

    Specifications

    Formula Weight 382.93
    Quantity 50mg
    Assay Percent Range >98%
    Molecular Formula C23H26N2O·HCl
    CAS 108050-82-4
    Synonym 3-[4-(3, 6-Dihydro-4-phenyl-1(2H)-pyridinyl)butyl]-1H-indol-5-ol hydrochloride
    Recommended Storage Desiccate at -20°C
    Chemical Name or Material Roxindole hydrochloride

    Desiccate at -20°C

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit