Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RP105/CD180 Mouse anti-Human, Allophycocyanin/Cy7, Clone: G28-8, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP262228APCCY7

Catalog No. N262228APC7


Add to cart

Description

Description

RP105/CD180 Monoclonal antibody specifically detects RP105/CD180 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

RP105/CD180
G28-8
Flow Cytometry
Conjugated, Purified
CD180
Human tonsillar B cells
Protein A purified
Adaptive Immunity, Immunology, Innate Immunity
Primary
4064.0
Human
Flow Cytometry
APC-Cyanine7
PBS with 0.05% Sodium Azide
CD180 antigenMGC126233, CD180 molecule, LY64MGC126234, Ly78, Lymphocyte antigen 64, lymphocyte antigen-64, radioprotective, 105kDa, Radioprotective 105 kDa protein, RP105lymphocyte antigen 64 (mouse) homolog, radioprotective, 105kD
Mouse
IgG1 Kappa
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
The antibody VI-01 reacts with vimentin, a 57 kDa intermediate filament expressed in variety of mesenchymal and mesodermal cell types. Cross-reactivity was found with smooth muscle desmin.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit