Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
RPA2 293T Cell Overexpression Lysate 1 (Denatured), Abnova
Manufacturer:  Abnova Corporation H00006118T01
Catalog No. 89-002-002
Description
Description
Western Blotting
Specifications
Specifications
Western Blot
293T
BC012157
6.8
Transient overexpression cell Lysate was tested with Anti-RPA2 antibody (H00006118-B01) by western blotting
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Human
RPA2 293T Cell Transient Overexpression Lysate(Denatured)
29.81
pCMV-RPA2 full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.