Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RPA2 Mouse anti-Canine, Human, Rat, Clone: OTI1H10, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA500765

Catalog No. 50-167-1908


Add to cart

Description

Description

Required for DNA recombination, repair and replication. The activity of RP-A is mediated by single-stranded DNA binding and protein interactions. Required for the efficient recruitment of the DNA double-strand break repair factor RAD51 to chromatin in response to DNA damage.
Specifications

Specifications

RPA2
Monoclonal
0.89 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P15927, Q63528
REPA2, RP-A p32, RP-A p34, RPA32
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
100856421, 59102, 6118
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation
OTI1H10
Unconjugated
RPA2
Liquid
RPA2
Full length human recombit protein of human RPA2 produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Canine, Human, Rat
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit