Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RPA2 Mouse anti-Human, Clone: OTI9A1, liquid, TrueMAB
Mouse Monoclonal Antibody
Manufacturer: OriGene TA500763S
Description
Required for DNA recombination, repair and replication. The activity of RP-A is mediated by single-stranded DNA binding and protein interactions. Required for the efficient recruitment of the DNA double-strand break repair factor RAD51 to chromatin in response to DNA damage.Specifications
RPA2 | |
Monoclonal | |
0.7 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P15927 | |
REPA2, RP-A p32, RP-A p34, RPA32 | |
Mouse | |
IgG1 | |
30 μL | |
-20° C, Avoid Freeze/Thaw Cycles | |
Primary | |
6118 |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation | |
OTI9A1 | |
Unconjugated | |
RPA2 | |
Liquid | |
RPA2 | |
Full length human recombit protein of human RPA2 produced in HEK293T cell. | |
Affinity Chromatography | |
RUO | |
Antibody | |
Monoclonal | |
Human |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title