Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RPA2 Mouse anti-Human, Clone: RPA2/4774, Novus Biologicals™
Mouse Monoclonal Antibody
$237.50 - $476.14
Specifications
Antigen | RPA2 |
---|---|
Clone | RPA2/4774 |
Concentration | 0.2 mg/ml |
Dilution | Western Blot 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array |
Applications | Western Blot, Immunohistochemistry (Paraffin), Peptide Array |
Description
RPA2 Monoclonal antibody specifically detects RPA2 in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin), Protein ArraySpecifications
RPA2 | |
0.2 mg/ml | |
Western Blot, Immunohistochemistry (Paraffin), Peptide Array | |
Unconjugated | |
Mouse | |
Cancer, Cell Cycle and Replication, Checkpoint signaling, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, Nucleotide Excision Repair, Stem Cell Markers | |
10 mM PBS with 0.05% BSA | |
REPA2, Replication factor A protein 2, replication protein A 32 kDa subunit, Replication protein A 34 kDa subunit, replication protein A2 (32kD), replication protein A2, 32kDa, RF-A protein 2, RP-A p32, RP-A p34, RPA32, RPA34 | |
Recombinant human full-length RPA2protein (Uniprot: P15927) | |
Primary | |
Store at 4°C. |
RPA2/4774 | |
Western Blot 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
Monoclonal | |
Purified | |
RUO | |
Human | |
P15927 | |
6118 | |
IgG1 κ | |
Protein A or G purified |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title