Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RPA2 Rabbit anti-Human, Clone: RPA2/3140R, Novus Biologicals™

Rabbit Monoclonal Antibody

$159.82 - $406.52

Specifications

Antigen RPA2
Clone RPA2/3140R
Concentration 0.2 mg/ml
Dilution Western Blot 1-2 ug/ml
Applications Western Blot
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122605
View Documents
Novus Biologicals
NBP30756610UG
100 μg
Each for $406.52
Add to cart
 
NB122606
View Documents
Novus Biologicals
NBP30756620UG
20 μg
Each for $159.82
Add to cart
 
Description

Description

RPA2 Monoclonal antibody specifically detects RPA2 in Human samples. It is validated for Western Blot
Specifications

Specifications

RPA2
0.2 mg/ml
Western Blot
Purified
RUO
Human
REPA2, Replication factor A protein 2, replication protein A 32 kDa subunit, Replication protein A 34 kDa subunit, replication protein A2 (32kD), replication protein A2, 32kDa, RF-A protein 2, RP-A p32, RP-A p34, RPA32, RPA34
Recombinant human full-length Replication protein A2 (RPA2) protein
Monoclonal
Protein A or G purified
RPA2/3140R
Western Blot 1-2 ug/ml
Unconjugated
Rabbit
Cancer, Cell Cycle and Replication, Checkpoint signaling, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, Nucleotide Excision Repair, Phospho Specific, Stem Cell Markers
10 mM PBS with 0.05% BSA
6118
IgG
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit