Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RPA70 Rabbit anti-Human, Clone: JB75-32, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP275648

Catalog No. NBP275648


Add to cart

Description

Description

RPA70 Monoclonal antibody specifically detects RPA70 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence.
Specifications

Specifications

RPA70
JB75-32
Unconjugated
TBS (pH7.4), 1%BSA, 40%Glycerol with 0.05% Sodium Azide
HSSB, MST075, MSTP075, REPA1RF-A protein 1, Replication factor A protein 1, replication protein A 70 kDa DNA-binding subunit, replication protein A1 (70kD), replication protein A1, 70kDa, RF-A, RP-A, RPA70RP-A p70, Single-stranded DNA-binding protein
Rabbit
IgG
Protein A purified
DNA Repair, Nucleotide Excision Repair
Primary
6117.0
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
1 mg/mL
Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500
Purified
RPA1
Recombinant protein corresponding to Human RPA70 aa 1-100.
68 kDa
100 μL
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit