Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RPAP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA559703
Description
Immunogen sequence: HITAVLTKII ERDTSSVAVN LPVPSGVAFP AVFLRSRDTQ GKSATSGKRS IFAQEIAARR IAEAKGPSVG EVVPNVGPPE GAVTCETPTP RNQGCQLPGS SHSFQGPNLV TGKGL Highest antigen sequence identity to the following orthologs: Mouse - 68%, Rat - 68%.
RNA polymerases transcribe nuclear genes for ribosomal RNA and are integral components of ribosomal biogenesis. RNA polymerase I (Pol I) is located in the nucleolus and transcribes class I genes, which code for large ribosomal RNA. RNA polymerase II (Pol II) synthesizes mRNA. RNA polymerase III (Pol III) transcribes class III genes, encoding a number of small ribosomal RNA molecules including tRNA and 5S rRNA. RPAP1 (RNA polymerase II associated protein 1) is a 1, 393 amino acid nuclear protein that is thought to connect Pol II to acetylated Histone H3 and regulators of protein complex formation. A member of the RPAP1 family, RPAP1 exists as three alternatively spliced isoforms that are encoded by a gene that maps to human chromosome 15q15.1.
Specifications
RPAP1 | |
Polyclonal | |
Unconjugated | |
RPAP1 | |
1190005L06Rik; A730023M06Rik; AU041788; AW107702; Kiaa1403; mKIAA1403; RNA polymerase II associated protein 1; RNA polymerase II-associated protein 1; RPAP1; RPAP-1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
26015 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
1.6 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BWH6 | |
RPAP1 | |
Recombinant protein corresponding to Human RPAP1. Recombinant protein control fragment (Product #RP-98046). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction