Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RPAP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5118246
This item is not returnable.
View return policy
Description
RPAP1 Polyclonal Antibody for Western Blot, IP
RNA polymerases transcribe nuclear genes for ribosomal RNA and are integral components of ribosomal biogenesis. RNA polymerase I (Pol I) is located in the nucleolus and transcribes class I genes, which code for large ribosomal RNA. RNA polymerase II (Pol II) synthesizes mRNA. RNA polymerase III (Pol III) transcribes class III genes, encoding a number of small ribosomal RNA molecules including tRNA and 5S rRNA. RPAP1 (RNA polymerase II associated protein 1) is a 1, 393 amino acid nuclear protein that is thought to connect Pol II to acetylated Histone H3 and regulators of protein complex formation. A member of the RPAP1 family, RPAP1 exists as three alternatively spliced isoforms that are encoded by a gene that maps to human chromosome 15q15.1.
Specifications
RPAP1 | |
Polyclonal | |
PBS with 0.03% ProClin 300; pH 7 | |
Q9BWH6 | |
RPAP1 | |
E. coli-derived Human RPAP1 fragment. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunoprecipitation, Western Blot | |
Unconjugated | |
RPAP1 | |
1190005L06Rik; A730023M06Rik; AU041788; AW107702; Kiaa1403; mKIAA1403; RNA polymerase II associated protein 1; RNA polymerase II-associated protein 1; RPAP1; RPAP-1 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
26015 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction