Learn More
RPAP1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 241161AP
Catalog No. 50-173-4240
Add to cart
Description
Description
RNA polymerases transcribe nuclear genes for ribosomal RNA and are integral components of ribosomal biogenesis. RNA polymerase I (Pol I) is located in the nucleolus and transcribes class I genes, which code for large ribosomal RNA. RNA polymerase II (Pol II) synthesizes mRNA. RNA polymerase III (Pol III) transcribes class III genes, encoding a number of small ribosomal RNA molecules including tRNA and 5S rRNA. RPAP1 (RNA polymerase II associated protein 1) is a 1, 393 amino acid nuclear protein that is thought to connect Pol II to acetylated Histone H3 and regulators of protein complex formation. A member of the RPAP1 family, RPAP1 exists as three alternatively spliced isoforms that are encoded by a gene that maps to human chromosome 15q15.1.
Specifications
Specifications
RPAP1
0.48 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q80TE0, Q9BWH6
KIAA1403, RPAP1
Rabbit
IgG
150 μL
-20°C
Primary
26015, 68925
Western Blot
Unconjugated
RPAP1
Liquid
RPAP1
RPAP1 Fusion Protein Ag7856
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse