Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RPIA Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5112699
Description
RPIA Polyclonal Antibody for Western Blot, IHC (P), ELISA
Defects in RPIA are the cause of ribose 5-phosphate isomerase deficiency. The exact function of RPIA remains unknown.
Specifications
RPIA | |
Polyclonal | |
Unconjugated | |
RPIA | |
Phosphoriboisomerase; ribose 5-phosphate epimerase; ribose 5-phosphate isomerase A; ribose 5-phosphate isomerase A (ribose 5-phosphate epimerase); ribose-5-phosphate isomerase; RPI; Rpia; RPIAD | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
19895, 22934 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
1.05 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P47968, P49247 | |
RPIA | |
Recombinant Human Ribose-5-phosphate isomerase protein (82-311AA). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction