Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RPIP8 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$106.19 - $352.89

Specifications

Antigen RPIP8
Dilution Western Blot 1.0 ug/ml
Applications Western Blot
Conjugate Unconjugated
Format Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB125293
View Documents
Novus Biologicals
NBP310196100UL
100 μg
Each for $352.89
Add to cart
 
NB125294
View Documents
Novus Biologicals
NBP31019625UL
25 μg
Each for $106.19
Add to cart
 
Description

Description

RPIP8 Polyclonal antibody specifically detects RPIP8 in Mouse samples. It is validated for Western Blot
Specifications

Specifications

RPIP8
Western Blot
Purified
RUO
Mouse
Rap2-interacting protein 8, RAP2IPRUN domain-containing protein 3A, RPIP-8, RPIP8RaP2 interacting protein 8, RUN domain containing 3A
The immunogen is a synthetic peptide directed towards the middle region of mouse RPIP8 (NP_001239276.1). Peptide sequence LKFTQSYDYLTDEEERHSAESSTSEDNSPEHPYLPLVTDEDSWYNKWHKM
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Western Blot 1.0 ug/ml
Unconjugated
Rabbit
Signal Transduction
PBS buffer, 2% sucrose
10900
Polyclonal
Affinity purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit