Learn More
RPL10 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 170131AP
Catalog No. 50-173-1328
Add to cart
Description
Description
RPL10 shares 98% amino acid sequence with RPL10L, so RPL10 antibody is able to recognize RPL10 and RPL10L protein.
This gene encodes a ribosomal protein that is a component of the 60S ribosome subunit. The related protein in chicken can bind to c-Jun and can repress c-Jun-mediated transcriptional activation. Some studies have detected an association between variation in this gene and autism spectrum disorders, though others do not detect this relationship. There are multiple pseudogenes of this gene dispersed throughout the genome. Alternative splicing results in multiple transcript variants.
Specifications
Specifications
RPL10
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P27635, P86048, Q6PDV7, Q6ZWV3, Q96L21
60S ribosomal protein L10, DXS648, DXS648E, Laminin receptor homolog, NOV, Protein QM, QM, ribosomal protein L10, RPL10, Tumor suppressor QM
Rabbit
IgG
150 μL
-20°C
Primary
110954, 140801, 238217, 299106, 6134, 81764
Western Blot
Unconjugated
RPL10
Liquid
RPL10, Rpl10l
RPL10 Fusion Protein Ag9969
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat