Learn More
Aviva Systems Biology Corp RPL10A Antibody - middle region (ARP56728_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP56728_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RPL10A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human RPL10A. Alternate names: Csa-19, NEDD6, L10AApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-62873