Learn More
RPL10A Mouse anti-Human, Clone: OTI4B2, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF809520
Catalog No. 50-166-9554
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 60S subunit. The protein belongs to the L1P family of ribosomal proteins. It is located in the cytoplasm. The expression of this gene is downregulated in the thymus by cyclosporin-A (CsA), an immunosuppressive drug. Studies in mice have shown that the expression of the ribosomal protein L10a gene is downregulated in neural precursor cells during development. This gene previously was referred to as NEDD6 (neural precursor cell expressed, developmentally downregulated 6), but it has been renamed RPL10A (ribosomal protein 10a). As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome.
Specifications
Specifications
RPL10A
Monoclonal
Unconjugated
RPL10A
Lyophilized
RPL10A
Full length human recombit protein of human RPL10A produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Immunohistochemistry (Paraffin), Western Blot
OTI4B2
PBS with 8% trehalose and no preservative; pH 7.3
P62906
CSA19, Csa-19, L10A, NEDD6
Mouse
IgG2a
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
4736