Learn More
RPL10A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 166811AP
Catalog No. 50-173-1109
Add to cart
Description
Description
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 60S subunit. The protein belongs to the L1P family of ribosomal proteins. It is located in the cytoplasm. The expression of this gene is downregulated in the thymus by cyclosporin-A (CsA), an immunosuppressive drug. Studies in mice have shown that the expression of the ribosomal protein L10a gene is downregulated in neural precursor cells during development. This gene previously was referred to as NEDD6 (neural precursor cell expressed, developmentally downregulated 6), but it has been renamed RPL10A (ribosomal protein 10a). As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome.
Specifications
Specifications
RPL10A
0.3 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P53026, P62906, P62907
60S ribosomal protein L10a, Csa 19, NEDD 6, NEDD6, ribosomal protein L10a, RPL10A
Rabbit
IgG
150 μL
-20°C
Primary
19896, 4736, 81729
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Unconjugated
RPL10A
Liquid
RPL10A
RPL10A Fusion Protein Ag10009
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat