Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RPL12 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5101382
Description
Antibody detects endogenous levels of total RPL12.
Eukaryotic translation initiation factor-5.
Specifications
RPL12 | |
Polyclonal | |
Unconjugated | |
RPL12 | |
60S ribosomal protein L12; E430018F03; L12; Large ribosomal subunit protein uL11; ribosomal protein L12; Rpl12 | |
Rabbit | |
Affinity chromatography | |
RUO | |
269261, 499782, 6136 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P23358, P30050, P35979 | |
RPL12 | |
A synthesized peptide derived from human RPL12, corresponding to a region within C-terminal amino acids. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction