Learn More
RPL15 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 167401AP
Catalog No. 50-173-1150
Add to cart
Description
Description
May play a role as co-chaperone of the Hsp70 family proteins HSPA1A, HSPA1B and HSPA8.
Specifications
Specifications
RPL15
0.23 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P61313, P61314, Q9CZM2
60S ribosomal protein L15, EC45, ribosomal protein L15, RPL10, RPL15, RPLY10, RPYL10
Rabbit
IgG
150 μL
-20°C
Primary
245981, 6138, 66480
Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
RPL15
Liquid
RPL15
RPL15 Fusion Protein Ag9948
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat