Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RPL17 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA551049
Description
RPL17 Polyclonal Antibody detects endogenous levels of RPL17 protein.
RPL17 Polyclonal antibody specifically detects RPL17 in Human, Mouse, Non-human Primate, Rat samples. It is validated for ELISA, Immunohistochemistry, Western Blot.Specifications
RPL17 | |
Polyclonal | |
Unconjugated | |
Rpl17 | |
60S ribosomal protein L17; 60S ribosomal protein L23; Amino acid starvation-induced protein; ASI; gene encoding putative NFkB activating protein; L17; L23; Large ribosomal subunit protein uL22; MGC117162; PD-1; ribosomal protein L17; RPL17; RPL23 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
291434, 319195, 6139 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P18621, P24049, Q9CPR4 | |
Rpl17 | |
Synthesized peptide derived from the Internal region of human Ribosomal Protein L17. | |
100 μL | |
Primary | |
Human, Mouse, Non-human Primate, Rat | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction