Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RPL26L1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA551050
Description
RPL26L1 Polyclonal Antibody detects endogenous levels of RPL26L1 protein.
RPL26L1 Polyclonal antibody specifically detects RPL26L1 in Human, Mouse, Rat samples. It is validated for ELISA, Immunohistochemistry, Western Blot.Specifications
RPL26L1 | |
Polyclonal | |
Unconjugated | |
RPL26L1 | |
60S ribosomal protein L26-like 1; Large ribosomal subunit protein uL24-like 1; ribosomal protein L26 homolog; ribosomal protein L26 like 1; ribosomal protein L26 pseudogene 1; RPL26L1; RPL26P1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
307636, 51121, 668405 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q9UNX3 | |
RPL26L1, Rpl26-ps3 | |
Synthesized peptide derived from the C-terminal region of human Ribosomal Protein L26L. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction