Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RPL27 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$436.00
Specifications
Antigen | RPL27 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP154629
|
Novus Biologicals
NBP154629 |
100 μL |
Each of 1 for $436.00
|
|
Description
RPL27 Polyclonal specifically detects RPL27 in Human samples. It is validated for Western Blot.Specifications
RPL27 | |
Polyclonal | |
Rabbit | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
ribosomal protein L27,60S ribosomal protein L27 | |
RPL27 | |
IgG | |
Affinity Purified | |
16 kDa |
Western Blot | |
Unconjugated | |
RUO | |
P61353 | |
6155 | |
Synthetic peptides corresponding to RPL27(ribosomal protein L27) The peptide sequence was selected from the middle region of RPL27. Peptide sequence SVDIPLDKTVVNKDVFRDPALKRKARREAKVKFEERYKTGKNKWFFQKLR. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title