Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RPL34 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA549958
Description
The antibody detects endogenous levels of total RPL34 protein.
RPL34 Polyclonal antibody specifically detects RPL34 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Western Blot.Specifications
RPL34 | |
Polyclonal | |
Unconjugated | |
RPL34 | |
1100001I22Rik; 60S ribosomal protein L34; L34; Large ribosomal subunit protein eL34; leukemia-associated protein; ribosomal protein L34; ribosomal protein L34-like 2; RL34; RPL34; Rpl34l2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
362041, 6164, 68436 | |
-20°C | |
Liquid |
Immunocytochemistry, Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P11250, P49207, Q9D1R9 | |
RPL34 | |
Synthesized peptide derived from internal of human RPL34. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction