Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RPL37 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5103546
Description
Antibody detects endogenous levels of total RPL37.
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 60S subunit. The protein belongs to the L37E family of ribosomal proteins. It is located in the cytoplasm. The protein contains a C2C2-type zinc finger-like motif.
Specifications
RPL37 | |
Polyclonal | |
Unconjugated | |
RPL37 | |
3110005M08Rik; 60S ribosomal protein L37; 60S ribosomal protein L37a; DKFZp686G1699; G1.16; L37; Large ribosomal subunit protein eL37; MGC99572; ribosomal protein L37; RPL37 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
6167, 67281, 81770 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P61927, P61928, Q9D823 | |
RPL37 | |
A synthesized peptide derived from human RPL37(Accession P61927), corresponding to amino acid residues C22-R72. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction