Learn More
RPL38 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 150551AP
Catalog No. 50-172-9937
Add to cart
Description
Description
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 60S subunit. The protein belongs to the L38E family of ribosomal proteins. It is located in the cytoplasm. Alternative splice variants have been identified, both encoding the same protein. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome, including one located in the promoter region of the type 1 angiotensin II receptor gene.
Specifications
Specifications
RPL38
0.15 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P63173, P63174, Q9JJI8
60S ribosomal protein L38, ribosomal protein L38, RPL38
Rabbit
IgG
150 μL
-20°C
Primary
6169, 67671, 689284
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
RPL38
Liquid
RPL38
RPL38 Fusion Protein Ag7038
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat