Learn More
RPL39 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 149901AP
Catalog No. 50-172-9892
Add to cart
Description
Description
Ribosomes, the organelles that catalyze protein synthesis, are composed of a small subunit (40S) and a large subunit (60S) that consist of over 80 distinct ribosomal proteins. Mammalian ribosomal proteins are encoded by multigene families that contain processed pseudogenes and one functional intron-containing gene within their coding regions. Ribosomal Protein L39 (RPL39), also known as 60S Ribosomal Protein L39, consists of 51 amino acids and is the smallest protein found in liver ribosomes. Ribosomal Protein L39 localizes to cytoplasm and is a member of the S39E family of ribosomal proteins. The gene encoding Ribosomal Protein L39 maps to human chromosome Xq24, and shares 92 percent homology with Ribosomal Protein L39L (Ribosomal Protein L39-like), which is also known as RPL39L, L39-2 or RPL39L1. Ribosomal Protein L39L is associated with the 60S ribosomal subunit and is expressed in testis with localization in nucleolus.
Specifications
Specifications
RPL39
0.2 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P62891, P62892, P62893
60S ribosomal protein L39, ribosomal protein L39, RPL39
Rabbit
IgG
150 μL
-20°C
Primary
25347, 6170, 67248
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
RPL39
Liquid
RPL39
RPL39 Fusion Protein Ag6975
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat