Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RPL7A, Rabbit anti-Human, Mouse, Clone: JE54-45, Invitrogen™

Rabbit Recombinant Monoclonal Antibody

Manufacturer:  Thermo Scientific MA536187

Catalog No. PIMA536187


Add to cart

Description

Description

RPL7A Recombinant Monoclonal antibody specifically detects RPL7A in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot
Specifications

Specifications

RPL7A
Recombinant Monoclonal
1 mg/mL
TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
P12970, P62424
60S ribosomal protein L7a; L7A; Large ribosomal subunit protein eL8; PLA-X polypeptide; ribosomal protein L7a; RPL7A; SURF3; Surf-3; surfeit 3; surfeit locus protein 3; thyroid hormone receptor uncoupling protein; TRUP
Recombinant protein within Human RPL7A.
Antigen affinity chromatography
RUO
Antibody
Monoclonal
Human, Mouse
Immunohistochemistry (Paraffin), Western Blot
JE54-45
Unconjugated
Rpl7a
Liquid
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
27176, 6130
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit