Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RPL7A Mouse anti-Human, Mouse, Clone: OTI3C5, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA527534

Catalog No. PIMA527534


Add to cart

Description

Description

Cytoplasmic ribosomes, organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 60S subunit. The protein belongs to the L7AE family of ribosomal proteins. It can interact with a subclass of nuclear hormone receptors, including thyroid hormone receptor, and inhibit their ability to transactivate by preventing their binding to their DNA response elements. This gene is included in the surfeit gene cluster, a group of very tightly linked genes that do not share sequence similarity. It is co-transcribed with the U24, U36a, U36b, and U36c small nucleolar RNA genes, which are located in its second, fifth, fourth, and sixth introns, respectively. This gene rearranges with the trk proto-oncogene to form the chimeric oncogene trk-2h, which encodes an oncoprotein consisting of the N terminus of ribosomal protein L7a fused to the receptor tyrosine kinase domain of trk. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome.
Specifications

Specifications

RPL7A
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
P12970, P62424
L7A, SURF3, TRUP, 60S ribosomal protein L7a, PLA-X polypeptide, surfeit 3, surfeit locus protein 3, thyroid hormone receptor uncoupling protein
Mouse
IgG2b
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
27176, 6130
Immunohistochemistry (Paraffin), Western Blot
OTI3C5
Unconjugated
RPL7A
Liquid
Rpl7a
Human recombit protein fragment corresponding to amino acids 2-266 of RPL7A produced in E.coli
Affinity Chromatography
RUO
Antibody
Monoclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit