Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RPLP1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA551140

Catalog No. PA551140


Add to cart

Description

Description

The antibody detects endogenous levels of total RPLP1 protein.

Plays an important role in the elongation step of protein synthesis.
Specifications

Specifications

RPLP1
Polyclonal
Unconjugated
RPLP1
Liquid
RPLP1
Fusion protein of human RPLP1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry, Western Blot
1.4 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
P05386
P1, LP1, RPP1
Rabbit
IgG
100 μL
-20°C
Primary
6176
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit