Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RPLP1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549548

Catalog No. PA549548

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with bovine, chicken, mouse and rat based on sequence homology.

Plays an important role in the elongation step of protein synthesis.
Specifications

Specifications

RPLP1
Polyclonal
PBS with 0.09% sodium azide
P05386
60S acidic ribosomal protein P1, RPLP1, RRP1
Rabbit
IgG
200 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
6176
Western Blot
Unconjugated
RPLP1
Liquid
RPLP1
KLH conjugated synthetic peptide between 18-49 amino acids from human RPLP1.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit