Learn More
Gibco™ RPMI 1640
Media with increased levels of vitamins
Manufacturer:  Gibco™ 670089
Catalog No. 67-008-9
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Contains increased levels of vitamins, the reducing agent glutathione, phenol red, 2 g/L glucose, 2 mM L-glutamine, and 2 g/L sodium bicarbonate. RPMI 1640 does not contain HEPES or sodium pyruvate.
Specifications
Specifications
Cell culture media
Basal media
Protect from light, 2°C to 8°C
500 mL