Learn More
BioworldSupplier Diversity Partner RPMI-1640 Medium w/ L-glutamine, w/o sodium bicarbonate, powder, cell culture tested, 50 L DFS Item
Manufacturer:  BioworldSupplier Diversity Partner 306110981
DescriptionRPMI 1640 Medium was developed at Roswell Park Memorial Institute in 1966 by Moore and his co-workers. A modification of McCoy's 5A Medium, it was formulated to support lymphoblastoid cells in suspension culture, but it has since been shown to support a wide variety of cells that are anchorage dependent. Originally intended to be used with a serum supplement, RPMI 1640 has been shown to support several cell lines in the absence of serum. It has also been widely used in fusion protocols and in the growth of hybrid cells.SpecificationsPowder formCell culture testedFormulated to contain 10.4 grams of powder per liter of medium.Supplement with 2.0 g/L sodium bicarbonate.Usage : bioWORLD's products are supplied for LABORATORY RESEARCH USE ONLY. The product may not be used as a drug, agricultural or pesticidal product , food additive or as a household chemical.
Catalog No. 50-254-215