Learn More
Ribophorin I Mouse anti-Canine, Human, Non-human primate, Clone: OTI3H3, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA525709
Catalog No. PIMA525709
Add to cart
Description
Description
Ribophorins 1 and 2 are abundant and highly conserved glycoproteins residing in the endoplasic reticulum, that participate in ribosome binding. Mammalian oligosaccharyltransferase activity is associated with a protein complex composed of RPN1, RPN2, and an oligosaccharyltransferase protein. RPN1 is a component of the proteasome base. The ubiquitin-like (UBL) domain of recombit Rad23 interacts with proteasomes through the leucine-rich repeat domain of RPN1. The RPN1 gene maps to chromosome 3 in somatic cell hybrids, and the RPN2 gene maps to chromosome 20 by in situ hybridization.
Specifications
Specifications
Ribophorin I
Monoclonal
0.75 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
E2RQ08, Q4R4T0, P04843
OST1, RBPH1, RPN-I, dolichyl-diphosphooligosaccharide--protein glycosyltransferase 67 kDa subunit, dolichyl-diphosphooligosaccharide--protein glycosyltransferase subunit 1, dolichyl-diphosphooligosaccharide-protein glycosyltransferase 67 kDa subunit, oligosaccharyltransfe
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
101925832, 476516, 6184
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
OTI3H3
Unconjugated
RPN1
Liquid
RPN1
Full length human recombit protein of RPN1 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Canine, Human, Non-human primate