Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RPP38 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548436

Catalog No. PA548436

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with bovine based on sequence homology.

Component of ribonuclease P, a protein complex that generates mature tRNA molecules by cleaving their 5 ft.-ends. RPP38 may associate transiently with RNase P RNA as a factor involved in the transport of H1 RNA to the putative site of its assembly in the cell, the nucleolus.
Specifications

Specifications

RPP38
Polyclonal
PBS with 0.09% sodium azide
P78345
Ribonuclease P protein subunit p38, RNaseP protein p38, RPP38
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
10557
Western Blot
Unconjugated
RPP38
Liquid
RPP38
KLH conjugated synthetic peptide between 248-277 amino acids from the C-terminal region of human RPP38.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit