Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RPRD1A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA598750
Description
RPRD1A Polyclonal Antibody for Western Blot, ELISA
P15RS is upregulated in cells overexpressing cyclin-dependent kinase inhibitor p15 and may have a role in cell cycle regulation (Liu et al., 2002 ).
Specifications
RPRD1A | |
Polyclonal | |
Unconjugated | |
RPRD1A | |
C77387; CDKN2B-related protein; cyclin-dependent kinase 2B-inhibitor-related protein; cyclin-dependent kinase inhibitor 2B-related protein; Cyclin-dependent kinase inhibitor 2B-related protein (p15INK4B-related protein); HsT3101; Kiaa4077; mKIAA4077; p15INK4B-related protein; P15RS; regulation of nuclear pre-mRNA domain containing 1A; regulation of nuclear pre-mRNA domain-containing protein 1A; RGD1307374; Rprd1a | |
Rabbit | |
Protein G | |
RUO | |
55197 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Western Blot | |
2 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q96P16 | |
RPRD1A | |
Recombinant Human Regulation of nuclear pre-mRNA domain-containing protein 1A protein (2-276AA). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction