Learn More
RPS15 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 149571AP
Catalog No. 50-172-9865
Add to cart
Description
Description
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 40S subunit. The protein belongs to the S19P family of ribosomal proteins. It is located in the cytoplasm. This gene has been found to be activated in various tumors, such as insulinomas, esophageal cancers, and colon cancers. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome. Alternative splicing results in multiple transcript variants.
Specifications
Specifications
RPS15
0.2 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P62841, P62843, P62845
40S ribosomal protein S15, ribosomal protein S15, RIG, RIG protein, RPS15
Rabbit
IgG
150 μL
-20°C
Primary
20054, 29285, 6209
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
RPS15
Liquid
RPS15
RPS15 Fusion Protein Ag6795
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat