Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RPS16 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP18002620UL

 View more versions of this product

Catalog No. NBP18002620


Add to cart

Description

Description

RPS16 Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications

Specifications

RPS16
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
Purified
RPS16
Synthetic peptide directed towards the middle region of human RPS16. Peptide sequence LVAYYQKYVDEASKKEIKDILIQYDRTLLVADPRRCKSKKFGGPGARAC.
44 kDa
20ul
Core ESC Like Genes, Stem Cell Markers
Primary
6217
Western Blot
Western Blot 1:1000
NP_001011
ribosomal protein S16,40S ribosomal protein S16
Rabbit
IgG
Protein A purified
RUO
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit