Learn More
RPS19 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 150851AP
Catalog No. 50-172-9965
Add to cart
Description
Description
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 40S subunit. The protein belongs to the S19E family of ribosomal proteins. It is located in the cytoplasm. Mutations in this gene cause Diamond-Blackfan anemia , a constitutional erythroblastopenia characterized by absent or decreased erythroid precursors, in a subset of patients. This suggests a possible extra-ribosomal function for this gene in erythropoietic differentiation and proliferation, in addition to its ribosomal function. Higher expression levels of this gene in some primary colon carcinomas compared to matched normal colon tissues has been observed. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome.
Specifications
Specifications
RPS19
0.31 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P17074, P39019, Q9CZX8
40S ribosomal protein S19, DBA, ribosomal protein S19, RPS19
Rabbit
IgG
150 μL
-20°C
Primary
108348115, 20085, 6223
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
RPS19
Liquid
RPS19
RPS19 Fusion Protein Ag7164
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat