Learn More
Invitrogen™ RPS2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA593178
Description
Immunogen sequence: MADDAGAAGG PGGPGGPGMG NRGGFRGGFG SGIRGRGRGR GRGRGRGRGA RGGKAEDKEW MPVTKLGRLV KDMKIKSLEE IYLFSLPIKE SEIIDFFLGA SLKDEVLKIM PVQKQTRAGQ RTRFKAFVAI GDYNGHVGLG VKCSKEVATA IRGAIILAKL SIVPVRRGYW GNKIGKPHTV PCKVTGRCGS VLVRLIPAPR GTGIVSAPVP KKLLMMAGID DCYTSARGCT ATLGNFAKAT FDAISKTYSY LTPDLWKETV FTKSPYQEFT DHLVKTHTRV SVQRTQAPAV ATT; Positive Samples: MCF7, HL-60, HeLa, 293T, THP-1, Mouse liver, Mouse brain, Rat liver, Rat brain.
Bacterial pathogens deliver type III effector proteins into the plant cell during infection. Plants express disease resistance (R) proteins that respond specifically to a particular type III effector by activating immune responses. Resistance proteins guard the plant against pathogens that contain an appropriate avirulence protein via an indirect interaction. RPS2 (resistance to Pseudomonas syringae protein 2) is a 909 amino acid plasma membrane protein with leucine-rich repeat, leucine zipper and P loop domains that confers resistance to Pseudomonas syringae infection by interacting with the avirulence gene AvrRpt2. Belonging to the disease resistance NB-LRR family, RPS2 directly associates with RIN4, which triggers plant resistance when RIN4 is degraded by AvrRpt2.
Specifications
RPS2 | |
Polyclonal | |
Unconjugated | |
Rps2 | |
40S ribosomal protein S2; 40S ribosomal protein S4; fc94f01; hypothetical protein LOC406565; ik:tdsubc_2h2; Llrep3; OK/KNS-cl.6; protein LLRep3; repeat family 3; ribosomal protein S2; ribosomal protein S2e; rps2; rps2e; Rps2r1; RPS4; S2; S4; Small ribosomal subunit protein uS5; tdsubc_2h2; wu:fb10e01; wu:fc94f01; wu:fj09b04; xx:tdsubc_2h2; zgc:85824 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
16898, 6187, 83789 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
0.47 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P15880, P25444, P27952 | |
Rps2 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-293 of human RPS2 (NP_0029432). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.